Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC14A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128588.3 | NP_001122060.3 | 11 | 1859 | ||
NM_001146036.2 | NP_001139508.2 | 11 | 1788 | |||
NM_001146037.1 | NP_001139509.1 | 9 | 1956 | |||
NM_001308278.1 | NP_001295207.1 | 9 | 1533 | |||
NM_001308279.1 | NP_001295208.1 | 8 | 1343 | |||
NM_015865.6 | NP_056949.4 | 10 | 1701 | |||
XM_005258333.1 | XP_005258390.1 | 7 | 1249 | |||
XM_006722526.3 | XP_006722589.1 | 11 | 1836 | |||
XM_011526142.1 | XP_011524444.1 | 10 | 2616 | |||
GenBank mRNA | AK091064.1 | - | 9 | 2184 | ||
AK123681.1 | - | 8 | 2760 | |||
AK127452.1 | - | 9 | 1445 | |||
L36121.1 | - | 8 | 1491 | |||
U35735.1 | - | 10 | 1637 |