Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023141.2 | NP_075630.2 | 6 | 1526 | ||
GenBank mRNA | AJ299404.1 | - | 6 | 1476 | ||
AK007814.1 | - | 6 | 1515 | |||
AK009693.1 | - | 6 | 1544 | |||
AK017908.1 | - | 6 | 1471 | |||
AK151048.1 | - | 6 | 1512 | |||
AK151390.1 | - | 7 | 1563 | |||
AK152243.1 | - | 6 | 1491 | |||
AK152358.1 | - | 6 | 1527 | |||
AK152484.1 | - | 6 | 1527 | |||
AK153391.1 | - | 6 | 1512 | |||
AK170296.1 | - | 6 | 1521 | |||
AK172355.1 | - | 6 | 1501 | |||
BC025525.1 | - | 6 | 1492 | |||
BC052851.1 | - | 6 | 1797 | |||
BC071216.1 | - | 6 | 1507 | 30439199 |