Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023320.2 | NP_075809.1 | 2 | 150 | ||
GenBank mRNA | AF168675.1 | - | 2 | 150 | ||
AK012945.1 | - | 2 | 311 | |||
AK014374.1 | - | 2 | 170 | |||
AK170025.1 | - | 2 | 265 | |||
AK170922.1 | - | 2 | 258 | |||
BC117003.1 | - | 2 | 157 | 40126322 | ||
BC125441.1 | - | 2 | 157 | 40129956 |