Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023324.2 | NP_075813.2 | 3 | 535 | ||
GenBank mRNA | AF302503.1 | - | 2 | 757 | ||
AK020915.1 | - | 3 | 569 | |||
AK029586.1 | - | 3 | 433 | |||
AK151660.1 | - | 3 | 536 | |||
AK152900.1 | - | 3 | 536 | |||
AK152997.1 | - | 3 | 536 | |||
AK153372.1 | - | 3 | 536 | |||
AK167525.1 | - | 3 | 536 | |||
BC016515.1 | - | 2 | 134 | 3600130 | ||
BC026695.1 | - | 3 | 499 |