Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agtpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048008.2 | NP_001041473.1 | 6 | 491 | ||
NM_001284218.1 | NP_001271147.1 | 6 | 616 | |||
NM_001284219.1 | NP_001271148.1 | 6 | 600 | |||
NM_001284221.1 | NP_001271150.1 | 5 | 366 | |||
NM_023328.3 | NP_075817.2 | 6 | 491 | |||
GenBank mRNA | AF219141.1 | - | 6 | 430 | ||
AK015394.1 | - | 5 | 330 | |||
AK051467.1 | - | 6 | 440 | |||
AK132695.1 | - | 6 | 431 | |||
AK133215.1 | - | 6 | 549 | |||
AK170046.1 | - | 6 | 434 | |||
BB614696.1 | - | 6 | 431 | |||
BC042475.1 | - | 6 | 600 | |||
BC060633.1 | - | 6 | 535 | |||
BC082335.1 | - | 5 | 354 | 30536788 | ||
CN527234.1 | - | 3 | 177 |