Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023464.2 | NP_075953.1 | 3 | 775 | ||
GenBank mRNA | AB041656.1 | - | 3 | 769 | ||
AK004482.1 | - | 1 | 654 | |||
AK088418.1 | - | 3 | 707 | |||
AK158412.1 | - | 890 | ||||
AK183655.1 | - | 1 | 324 | |||
BC023033.1 | - | 3 | 738 | |||
BC037081.1 | - | 3 | 752 | 5098653 | ||
BC048360.1 | - | 3 | 754 | 5067090 |