Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nuf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023284.3 | NP_075773.2 | 4 | 406 | ||
GenBank mRNA | AF326732.1 | - | 5 | 447 | ||
AK010351.1 | - | 4 | 328 | |||
AK012011.1 | - | 4 | 340 | |||
AK089939.1 | - | 5 | 434 | |||
AK132467.1 | - | 4 | 407 | |||
AK162249.1 | - | 5 | 442 | |||
BC020026.1 | - | 5 | 415 | 3587655 |