Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023320.2 | NP_075809.1 | 2, 3 | 221 | ||
GenBank mRNA | AF168675.1 | - | 2, 3 | 221 | ||
AK012945.1 | - | 2, 3 | 382 | |||
AK014374.1 | - | 2, 3 | 241 | |||
AK170025.1 | - | 2, 3 | 336 | |||
AK170922.1 | - | 2, 3 | 329 | |||
BC117003.1 | - | 2, 3 | 228 | 40126322 | ||
BC125441.1 | - | 2, 3 | 228 | 40129956 |