Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023530.2 | NP_076019.2 | 1 | 269 | ||
GenBank mRNA | AF339738.1 | - | 1 | 291 | ||
AK008035.1 | - | 1 | 295 | |||
AY358033.1 | - | 1 | 298 | |||
BC021592.1 | - | 1 | 269 | 5133141 |