Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023500.2 | NP_075989.1 | 3 | 2753 | ||
GenBank mRNA | AF155511.1 | - | 3 | 2690 | ||
AY534248.1 | - | 3 | 2735 |