Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023580.4 | NP_076069.2 | 1, 2 | 147 | ||
GenBank mRNA | AF131197.1 | - | 1, 2 | 89 | ||
AK017662.1 | - | 1, 2 | 115 | |||
AK028478.1 | - | 1, 2 | 148 | |||
BC071215.1 | - | 1, 2 | 111 | 4196138 |