Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023603.3 | NP_076092.1 | 1, 2 | 928 | ||
GenBank mRNA | AF272847.1 | - | 1, 2 | 221 | ||
AK157783.1 | - | 2, 3 | 565 | |||
AY034062.1 | - | 1, 2 | 843 | |||
BC080848.1 | - | 1, 2 | 894 | |||
BC089305.1 | - | 1, 2 | 894 | 30360144 |