Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256130.1 | NP_001243059.1 | 3 | 589 | ||
NM_001256131.1 | NP_001243060.1 | 2 | 468 | |||
NM_001256132.1 | NP_001243061.1 | 3 | 393 | |||
NM_023647.6 | NP_076136.2 | 3 | 1343 | |||
GenBank mRNA | AB041581.1 | - | 3 | 526 | ||
AK008106.1 | - | 3 | 575 | |||
AK011248.1 | - | 2 | 415 | |||
AK012859.1 | - | 3 | 527 | |||
AK028734.1 | - | 3 | 1301 | |||
AK031255.1 | - | 3 | 554 | |||
AK041427.1 | - | 3 | 590 | |||
AK076051.1 | - | 3 | 546 | |||
AK076184.1 | - | 3 | 529 | |||
AK154755.1 | - | 3 | 547 | |||
AK183552.1 | - | 2 | 299 | |||
AK196701.1 | - | 3 | 315 | |||
BC031109.1 | - | 1 | 412 | |||
BC038499.1 | - | 3 | 531 | 4482150 |