Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023668.2 | NP_076157.2 | 3 | 333 | ||
GenBank mRNA | AF290472.1 | - | 3 | 265 | ||
AF323918.1 | - | 3 | 353 | |||
AK011168.1 | - | 3 | 243 | |||
AK146029.1 | - | 3 | 262 | |||
AK212133.1 | - | 2 | 112 | |||
BC021434.1 | - | 229 | 5029832 | |||
BC046796.1 | - | 3 | 264 | 5718032 | ||
CJ092278.1 | - | 3 | 334 |