Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023670.3 | NP_076159.3 | 1, 2 | 666 | ||
GenBank mRNA | AB046173.1 | - | 1, 2 | 183 | ||
AK011689.1 | - | 1, 2 | 447 | |||
AK011797.1 | - | 1, 2 | 417 | |||
AK088465.1 | - | 1, 2 | 425 | |||
AK147140.1 | - | 1, 2 | 670 | |||
BC045138.1 | - | 1, 2 | 296 | 6306448 | ||
BC049082.1 | - | 1, 2 | 332 | 5706238 |