Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crybb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023695.2 | NP_076184.1 | 4, 5 | 468 | ||
GenBank mRNA | AF106853.1 | - | 4, 5 | 468 | ||
AK014012.1 | - | 4, 5 | 470 | |||
AK053869.1 | - | 4, 5 | 469 | |||
BC029012.1 | - | 3, 4 | 452 | 5358714 |