Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930550L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023774.3 | NP_076263.1 | 2 | 419 | ||
GenBank mRNA | AF319984.1 | - | 1 | 269 | ||
AK016090.1 | - | 2 | 397 | |||
BC087920.1 | - | 2 | 419 | 30281980 |