Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mvk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF137598.1 | - | 6 | 682 | ||
AK009261.1 | - | 8 | 792 | |||
AK031084.1 | - | 6 | 1462 | |||
AK149630.1 | - | 7 | 779 | |||
BC005606.1 | - | 7 | 735 | 3498140 |