Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081490.2 | NP_001074959.1 | 4 | 345 | ||
NM_023605.2 | NP_076094.2 | 4 | 647 | |||
GenBank mRNA | AB041585.1 | - | 4 | 616 | ||
AK018482.1 | - | 4 | 598 | |||
AK077607.1 | - | 4 | 308 | |||
AK182638.1 | - | 3 | 237 | |||
BC020074.1 | - | 4 | 618 | 3710611 | ||
DV657928.1 | - | 4 | 333 |