Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ern1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023913.2 | NP_076402.1 | 4, 5 | 408 | ||
GenBank mRNA | AB031332.1 | - | 4, 5 | 408 | ||
AK018505.1 | - | 4, 5 | 449 | |||
AK077041.1 | - | 427 | ||||
AK170988.1 | - | 4, 5 | 437 |