Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxct1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024188.6 | NP_077150.1 | 5 | 854 | ||
GenBank mRNA | AB085609.1 | - | 5 | 596 | ||
AK009027.1 | - | 5 | 634 | |||
AK010790.1 | - | 5 | 597 | |||
AK011354.1 | - | 5 | 534 | |||
AK078086.1 | - | 5 | 854 | |||
AK146158.1 | - | 5 | 612 | |||
AK146342.1 | - | 5 | 599 | |||
AK151662.1 | - | 5 | 593 | |||
AK151695.1 | - | 5 | 584 | |||
AK167681.1 | - | 5 | 684 | |||
AK181182.1 | - | 1 | 70 | |||
AK181314.1 | - | 1 | 69 | |||
AK184597.1 | - | 1 | 70 | |||
BC003422.1 | - | 5 | 558 | 2598824 | ||
EF095212.1 | - | 2 | 247 |