Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168658.1 | NP_001162129.1 | 3 | 314 | ||
NM_001168659.1 | NP_001162130.1 | 3 | 314 | |||
NM_024201.3 | NP_077163.1 | 3 | 308 | |||
GenBank mRNA | AK002564.1 | - | 3 | 296 | ||
AK007738.1 | - | 3 | 260 | |||
AK164539.1 | - | 3 | 315 | |||
AK170932.1 | - | 3 | 301 | |||
BC003345.1 | - | 3 | 288 | 3257525 | ||
BC049099.1 | - | 3 | 266 | 5721356 |