Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024211.3 | NP_077173.1 | 2 | 411 | ||
GenBank mRNA | AI931249.1 | - | 2 | 372 | ||
AK003957.1 | - | 1 | 50 | |||
AK009487.1 | - | 2 | 357 | |||
AK009824.1 | - | 2 | 311 | |||
AK168951.1 | - | 2 | 336 | |||
AK181540.1 | - | 1 | 93 | |||
AK187524.1 | - | 1 | 93 | |||
AK202646.1 | - | 1 | 93 | |||
AK212146.1 | - | 1 | 83 | |||
AK213879.1 | - | 1 | 93 | |||
AK215545.1 | - | 1 | 83 | |||
BC003455.1 | - | 2 | 387 | 2648336 | ||
BC019631.1 | - | 2 | 365 | 4023179 | ||
BY006289.1 | - | 2 | 410 |