Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143776.1 | NP_001137248.1 | 3 | 304 | ||
NM_001143777.1 | NP_001137249.1 | 4 | 408 | |||
NM_024244.4 | NP_077206.3 | 3 | 304 | |||
GenBank mRNA | AK004797.1 | - | 3 | 288 | ||
AK021133.1 | - | 4 | 247 | |||
AK032053.1 | - | 4 | 220 | |||
AK036299.1 | - | 4 | 371 | |||
AK039513.1 | - | 4 | 372 | |||
AK041147.1 | - | 3 | 305 | |||
AK044049.1 | - | 4 | 331 | |||
AK081939.1 | - | 4 | 409 | |||
AK082202.1 | - | 3 | 300 | |||
AK220421.1 | - | 4 | 244 | |||
BB637603.1 | - | 3 | 306 | |||
BC129839.1 | - | 3 | 260 | 40060810 | ||
BC129840.1 | - | 3 | 260 | 40060811 |