Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Set | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204875.1 | NP_001191804.1 | 6 | 973 | ||
NM_023871.4 | NP_076360.1 | 6 | 838 | |||
GenBank mRNA | AB044937.1 | - | 6 | 722 | ||
AK019960.1 | - | 6 | 722 | |||
AK172327.1 | - | 6 | 982 | |||
BC018255.1 | - | 6 | 791 | 4014298 | ||
BC103791.1 | - | 6 | 666 | 6511600 |