Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuedc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164290.1 | NP_001157762.1 | 4, 5 | 438 | ||
NM_001164291.1 | NP_001157763.1 | 3, 4 | 330 | |||
NM_001164292.1 | NP_001157764.1 | 3, 4 | 327 | |||
NM_001164293.1 | NP_001157765.1 | 3, 4 | 330 | |||
NM_001164294.1 | NP_001157766.1 | 3, 4 | 330 | |||
NM_001164295.1 | NP_001157767.1 | 3, 4 | 327 | |||
NM_024192.2 | NP_077154.2 | 4, 5 | 429 | |||
GenBank mRNA | AB232359.1 | - | 2, 3 | 228 | ||
AK003457.1 | - | 3, 4 | 285 | |||
AK008842.1 | - | 3, 4 | 293 | |||
AK013884.1 | - | 3, 4 | 288 | |||
AK186231.1 | - | 3, 4 | 253 | |||
AK207375.1 | - | 4, 5 | 355 | |||
BC003442.1 | - | 4, 5 | 393 | 2650719 | ||
BY279028.1 | - | 3, 4 | 329 | |||
BY312150.1 | - | 3, 4 | 291 | |||
CK625575.1 | - | 3, 4 | 256 | |||
CX218143.1 | - | 4, 5 | 368 | |||
CX223880.1 | - | 4, 5 | 394 |