Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199188.1 | NP_001186117.1 | 5 | 582 | ||
NM_024225.5 | NP_077187.1 | 4 | 493 | |||
GenBank mRNA | AK003091.1 | - | 206 | |||
AK007676.1 | - | 4 | 441 | |||
AK029463.1 | - | 4 | 453 | |||
AK031873.1 | - | 5 | 474 | |||
AK049129.1 | - | 5 | 474 | |||
AK078776.1 | - | 5 | 475 | |||
AK144710.1 | - | 5 | 475 | |||
AK152311.1 | - | 5 | 518 | |||
AK159115.1 | - | 1 | 36 | |||
AK159904.1 | - | 4 | 398 | |||
AK167362.1 | - | 4 | 413 | |||
AK169545.1 | - | 4 | 714 | |||
AK169786.1 | - | 5 | 474 | |||
AK177366.1 | - | 3 | 307 | |||
AK184438.1 | - | 3 | 306 | |||
AK187575.1 | - | 3 | 305 | |||
AK214265.1 | - | 3 | 306 | |||
BC002242.1 | - | 5 | 551 | 3492262 |