Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGXT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000030.2 | NP_000021.1 | 1, 2 | 282 | ||
GenBank mRNA | AB292648.1 | - | 1, 2 | 183 | ||
AF348451.1 | - | 1, 2 | 73 | |||
AK292754.1 | - | 1, 2 | 203 | |||
AK298400.1 | - | 1, 2 | 68 | |||
AK298420.1 | - | 1, 2 | 198 | |||
AK315331.1 | - | 1, 2 | 230 | |||
AK316495.1 | - | 1, 2 | 68 | |||
BC132819.1 | - | 1, 2 | 279 | 40146841 | ||
D13368.1 | - | 1, 2 | 269 | |||
X53414.1 | - | 1, 2 | 283 | |||
X56092.1 | - | 1, 2 | 182 |