Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Riok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024242.3 | NP_077204.2 | 16, 17 | 2073 | ||
GenBank mRNA | AK014182.1 | - | 3, 4 | 800 | ||
AK152707.1 | - | 16, 17 | 1822 | |||
AK161315.1 | - | 17, 18 | 2316 | |||
BC002158.1 | - | 10, 11 | 1030 | 3485522 |