Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timmdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024273.2 | NP_077235.2 | 2 | 414 | ||
GenBank mRNA | AK012774.1 | - | 2 | 388 | ||
AK015460.1 | - | 2 | 392 | |||
AK081781.1 | - | 2 | 357 | |||
BC002243.1 | - | 2 | 376 | 3492342 | ||
CJ056343.1 | - | 2 | 416 |