Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000035.3 | NP_000026.2 | 3 | 227 | ||
GenBank mRNA | AK290795.1 | - | 3 | 221 | ||
BC029399.1 | - | 3 | 221 | 4593670 | ||
X01098.1 | - | 3 | 211 | |||
X02747.1 | - | 3 | 270 |