Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024442.1 | NP_077762.1 | 3 | 335 | ||
GenBank mRNA | AF233646.1 | - | 3 | 335 | ||
AK078557.1 | - | 3 | 396 | |||
AK146232.1 | - | 3 | 408 | |||
BC026539.1 | - | 3 | 367 | 4982322 |
Gene Symbol: Cyp4f37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100187.1 | NP_001093657.1 | 3 | 343 | ||
GenBank mRNA | AK132492.1 | - | 3 | 343 | ||
BC151030.1 | - | 328 | 9087943 |