Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024479.2 | NP_077799.2 | 2, 3 | 272 | ||
GenBank mRNA | AK042578.1 | - | 2, 3 | 163 | ||
AK081683.1 | - | 2, 3 | 291 | |||
AK132481.1 | - | 2, 3 | 261 | |||
AY138964.1 | - | 2, 3 | 272 | |||
AY354925.2 | - | 2, 3 | 272 | |||
AY354926.1 | - | 2, 3 | 173 | |||
AY354927.1 | - | 2, 3 | 272 | |||
BC002286.1 | - | 2, 3 | 263 | 3589835 |