Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ278264.1 | - | 6 | 541 | ||
AK154786.1 | - | 6 | 567 | |||
AK170037.1 | - | 6 | 642 | |||
AY165625.1 | - | 6 | 556 | |||
BC106828.1 | - | 6 | 552 | 40044453 | ||
BC106829.1 | - | 6 | 556 | 40044455 | ||
BG077775.2 | - | 6 | 542 |