Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ky | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024291.3 | NP_077253.3 | 2, 3 | 211 | ||
GenBank mRNA | AJ293727.2 | - | 2, 3 | 233 | ||
AK047076.1 | - | 2, 3 | 602 |