Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024262.1 | NP_077224.1 | 1 | 1491 | ||
GenBank mRNA | AK004714.1 | - | 1 | 1484 | ||
AK031106.1 | - | 1 | 1465 | |||
AK035089.1 | - | 1 | 458 | |||
BC002247.1 | - | 1 | 96 | |||
BC020005.1 | - | 1 | 1491 | 3981459 |