Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000073.2 | NP_000064.1 | 2, 3 | 167 | ||
XM_005271724.3 | XP_005271781.1 | 2, 3 | 204 | |||
XM_006718941.2 | XP_006719004.1 | 2, 3 | 155 | |||
GenBank mRNA | AK223314.1 | - | 2, 3 | 120 | ||
AK313966.1 | - | 2, 3 | 155 | |||
AK315895.1 | - | 2, 3 | 111 | |||
BC113830.1 | - | 2, 3 | 113 | 40029587 | ||
X04145.1 | - | 2, 3 | 124 |