Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TAZ SIRNA Assays ›
Gene Symbol:
TAZ
Gene Name:
tafazzin
Gene Aliases:
BTHS, CMD3A, EFE, EFE2, G4.5, LVNCX, Taz1
Chromosome Location:
Chr.X: 154411406 - 154421726 on Build GRCh38
UniGene ID:
Hs.409911
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
7811
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TAZ
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000116.4
NP_000107.1
4
652
NM_001303465.1
NP_001290394.1
4
706
NM_181311.3
NP_851828.1
4
652
NM_181312.3
NP_851829.1
4
652
NM_181313.3
NP_851830.1
4
652
NR_024048.2
4
758
XM_006724836.1
XP_006724899.1
4
683
XM_006724837.1
XP_006724900.1
4
683
XM_006724839.1
XP_006724902.1
4
683
XM_006724841.3
XP_006724904.1
4
404
XM_006724842.3
XP_006724905.1
4
404
XM_011531189.1
XP_011529491.1
4
683
XM_011531191.2
XP_011529493.1
3
842
XM_017029761.1
XP_016885250.1
4
629
XM_017029762.1
XP_016885251.1
4
683
XM_017029763.1
XP_016885252.1
4
629
XM_017029764.1
XP_016885253.1
2
181
XM_017029765.1
XP_016885254.1
4
851
GenBank mRNA
AK291848.1
-
4
520
AL531401.3
-
4
739
AY231461.1
-
4
334
AY231462.1
-
4
334
AY231463.1
-
4
334
AY231464.1
-
4
334
AY258036.1
-
4
334
AY258037.1
-
4
334
AY258038.1
-
4
334
AY258039.1
-
4
334
BC005062.1
-
4
715
BC011515.1
-
3
348
4339983
BX401320.2
-
4
492
CA488175.1
-
3
248
DQ884408.1
-
4
380
KU178348.1
-
3
251
KU178349.1
-
4
326
KU178350.1
-
4
326
KU178351.1
-
4
413
KU178352.1
-
4
326
KU178353.1
-
4
326
X74607.1
-
1
35
X87198.1
-
1
35
X92762.1
-
4
614
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
heart development
skeletal muscle tissue development
hemopoiesis
cardiolipin biosynthetic process
mitochondrial respiratory chain complex I assembly
cardiolipin acyl-chain remodeling
cristae formation
mitochondrial ATP synthesis coupled electron transport
cardiac muscle tissue development
cardiac muscle contraction
1-acylglycerol-3-phosphate O-acyltransferase activity
O-acyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0