Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025418.3 | NP_079694.2 | 8 | 1074 | ||
GenBank mRNA | AK002611.1 | - | 8 | 1073 | ||
AK003213.1 | - | 7 | 991 | |||
AK004323.1 | - | 8 | 1145 | |||
AK009227.2 | - | 8 | 1063 | |||
AK042917.1 | - | 2 | 309 | |||
BC026752.1 | - | 8 | 1067 | 4034998 |