Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nenf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025424.2 | NP_079700.1 | 1, 2 | 211 | ||
GenBank mRNA | AB126218.1 | - | 1, 2 | 185 | ||
AK002413.1 | - | 1, 2 | 212 | |||
AK004329.1 | - | 1, 2 | 200 | |||
AK009304.1 | - | 1, 2 | 202 | |||
AK020403.1 | - | 1, 2 | 193 | |||
AK155319.1 | - | 1, 2 | 210 | |||
BC048464.1 | - | 1, 2 | 199 | 6596336 |