Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025473.3 | NP_079749.1 | 4 | 618 | ||
GenBank mRNA | AK007715.1 | - | 4 | 619 | ||
BC011237.1 | - | 4 | 616 | |||
BC019752.1 | - | 5 | 384 | |||
BC054411.1 | - | 4 | 290 | |||
BC069871.1 | - | 4 | 753 | 6467541 |