Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025520.3 | NP_079796.1 | 5 | 301 | ||
GenBank mRNA | AK008480.1 | - | 4 | 559 | ||
AK009617.1 | - | 5 | 295 | |||
AK011218.1 | - | 5 | 287 | |||
BC048459.1 | - | 5 | 276 | 6601124 | ||
BC061085.1 | - | 285 | 30295556 | |||
BC132626.1 | - | 5 | 301 | 40130903 | ||
BC132628.1 | - | 5 | 301 | 40130905 | ||
CB204385.1 | - | 5 | 294 |