Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163536.1 | NP_001157008.1 | 5 | 692 | ||
NM_001163537.1 | NP_001157009.1 | 5 | 712 | |||
NM_025489.3 | NP_079765.3 | 5 | 699 | |||
GenBank mRNA | AK005725.1 | - | 5 | 679 | ||
AK005749.1 | - | 5 | 668 | |||
BC049579.1 | - | 5 | 642 | 6772915 | ||
CA463124.1 | - | 3 | 413 |