Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025509.3 | NP_079785.1 | 3 | 427 | ||
GenBank mRNA | AK009237.1 | - | 3 | 402 | ||
AK011085.1 | - | 3 | 379 | |||
AK013011.1 | - | 3 | 386 | |||
AK135332.1 | - | 3 | 383 | |||
BC014799.1 | - | 3 | 402 | |||
BC021935.1 | - | 3 | 401 | 5149713 |