Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025541.3 | NP_079817.1 | 1, 2 | 333 | ||
GenBank mRNA | AK007804.1 | - | 1, 2 | 189 | ||
AK010210.1 | - | 1, 2 | 343 | |||
AK012376.1 | - | 1, 2 | 324 | |||
AK031513.1 | - | 1, 2 | 299 | |||
AK088618.1 | - | 1, 2 | 276 | |||
BC027628.1 | - | 1, 2 | 125 | 1178866 |