Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025580.2 | NP_079856.2 | 3 | 342 | ||
GenBank mRNA | AF318058.1 | - | 3 | 446 | ||
AK009545.1 | - | 3 | 319 | |||
AK012976.1 | - | 3 | 321 | |||
AK166548.1 | - | 3 | 295 | |||
AK179109.1 | - | 2 | 243 | |||
AK181664.1 | - | 2 | 242 | |||
AY039041.1 | - | 3 | 305 | |||
AY299972.1 | - | 3 | 305 | |||
BC008274.1 | - | 3 | 351 | |||
BC058945.1 | - | 3 | 321 | 6812127 | ||
BC116236.1 | - | 3 | 429 | |||
BC116237.1 | - | 3 | 429 | |||
CK623847.1 | - | 3 | 312 |