Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025611.5 | NP_079887.3 | 22 | 4517 | ||
GenBank mRNA | AK010907.1 | - | 1 | 44 | ||
AK147156.1 | - | 22 | 4275 | |||
AK147905.1 | - | 22 | 4236 | |||
AK170808.1 | - | 22 | 4290 | |||
AK172882.1 | - | 21 | 3590 | |||
AK200068.1 | - | 2 | 238 | |||
BC019645.1 | - | 12 | 1805 | |||
BC024345.1 | - | 1 | 309 | |||
BC026946.1 | - | 4 | 625 | |||
BC059865.1 | - | 18 | 3033 | |||
BC065096.1 | - | 21 | 4004 | |||
BC100544.1 | - | 1 | 153 | |||
BC138998.1 | - | 22 | 4455 | 8862020 | ||
BC139000.1 | - | 22 | 4455 | 8862022 |