Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025597.2 | NP_079873.1 | 3 | 373 | ||
GenBank mRNA | AK007930.1 | - | 3 | 372 | ||
AK009008.1 | - | 3 | 335 | |||
AK012320.1 | - | 3 | 374 | |||
AK013608.1 | - | 3 | 359 | |||
AK138123.1 | - | 3 | 371 | |||
BC028669.1 | - | 3 | 529 | |||
BC036989.1 | - | 3 | 370 | 4989233 |