Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110809.2 | NP_001104279.1 | 3 | 229 | ||
NM_001286857.1 | NP_001273786.1 | 3 | 843 | |||
NM_025663.4 | NP_079939.3 | 4 | 364 | |||
GenBank mRNA | AK011617.1 | - | 3 | 228 | ||
AK017310.1 | - | 3 | 179 | |||
AK018306.1 | - | 3 | 203 | |||
AK078828.1 | - | 3 | 227 | |||
AK169111.1 | - | 3 | 218 | |||
BC002265.1 | - | 3 | 200 | |||
BC019490.1 | - | 4 | 346 | 4039649 | ||
BC080676.1 | - | 3 | 821 | |||
BI658293.1 | - | 4 | 364 |