Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025675.4 | NP_079951.1 | 3 | 313 | ||
GenBank mRNA | AK006965.1 | - | 3 | 271 | ||
AK007021.1 | - | 3 | 316 | |||
AK017595.1 | - | 3 | 269 | |||
AK033888.1 | - | 3 | 278 | |||
AK036021.1 | - | 3 | 349 | |||
AK082013.1 | - | 3 | 350 | |||
AK088160.1 | - | 3 | 315 | |||
AK145789.1 | - | 3 | 273 | |||
BC022995.1 | - | 3 | 338 | 5363234 |